Cleanroom Overalls, CleanMax®
Product Details & Documents
Clean manufactured disposable coveralls are individually packaged and folded to promote easier gowning with less excursions.
- Clean manufactured in silicone-free environment
- Latex-free
- Tunnelled elastic on wrists with thumb loops
- Zipper closure
- For critical environments
- Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
Specifikationer
- Tøj størrelse:XXL
- Farve:hvid
- Beskrivelse:Clean manufactured coverall
- Seq:04
- Cat. No.:114-9584
- Renrum klasse:100/ISO 5
- Outfit applikationer:Cleanroom
- Lukkestørrelse:Med lynlås