Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Threaded fittings

Socket, reduced, inner, NW 17,5 mm, PP, 12,7 - 19,05 mm
Katalog # BURK8502-1234
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8502-1234
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Socket, reduced, inner, NW 17,5 mm, PP, 12,7 - 19,05 mm

Price
67,20 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    12,7 - 19,05 mm
  • Beskrivelse:
    Socket, reduced, inner, NW 17,5 mm
  • Materiale:
    PP