Orders LinkContactUsLinkComponent
GloCell™ Fixable Viability Dye UV 450
GloCell™ Fixable Viability Dye UV 450
GloCell™ Fixable viability dye UV 450, 100 Tests
Katalog # STMC75008.1
Marketsource
Leverandør:   Stemcell Technologies
GloCell™ Fixable Viability Dye UV 450
Katalog # STMC75008.1
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
GloCell™ Fixable Viability Dye UV 450
GloCell™ Fixable Viability Dye UV 450
GloCell™ Fixable viability dye UV 450, 100 Tests
CAS nummer:  
Price
967,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Amine-labelling fluorescent dye for live/dead staining of mammalian cells - 100 tests.
GloCell™ Fixable viability dye UV 450 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be utilised prior to fixation, permeabilisation, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay.
GloCell™ Fixable viability dye UV 450 is excited by the UV laser at 355 nm and it has a fluorescence emission maximum of 450 nm that can be detected using the 450/50 band pass filter used to detect DAPI, Hoechst 33258, etc.
CAS No: 67-68-5
Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Pakn:
    100 Tests
  • Beskrivelse:
    GloCell™ Fixable viability dye UV 450