Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant GDNF

Human Recombinant GDNF

500 µG
Katalog # STMC78058.3
Marketsource
Leverandør:   Stemcell Technologies
Human Recombinant GDNF
Katalog # STMC78058.3
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Human Recombinant GDNF
Human Recombinant GDNF

500 µG

CAS nummer:  
Price
21.439,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Glial cell line-derived neurotrophic factor (GDNF) is a neurotrophic factor and a member of the tumor growth factor (TGF)-beta superfamily. The GDNF family of growth factors also includes neurturin, persephin, and artemin, which have seven conserved cysteine residues called cysteine-knots (Treanor et al.). GDNF family ligands signal through binding to specific GDNF-family receptor-α (GFRα) co-receptors and activate the RET receptor tyrosine kinase (Durbec et al.). Four different forms of GFRα co-receptors have been characterized (GFRα 1-4); GDNF binds specifically to GFRα1 prior to forming a complex with RET (Airaksinen and Saarma). GDNF is known to promote survival and morphological differentiation of midbrain dopaminergic neurons in both in vivo and in vitro studies and increases their high-affinity dopamine uptake (Granholm et al.; Lin et al.). GDNF has also been shown to have restorative effects on dying dopaminergic neurons in response to degenerative toxins (Aoi et al.). GDNF, together with Human Recombinant BDNF (brain-derived neurotrophic factor), BrainPhys™ Neuronal Medium, and other supplements, can be used to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into neurons (Bardy et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    500 µG
  • Cat. No.:
    STMC78058.3
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    GDNF
  • Renhed:
    ≥95%
  • Species:
    Human

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.