Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant TGF-beta 1

5 µG
Katalog # STMC78067
Marketsource
Leverandør:   Stemcell Technologies
Human Recombinant TGF-beta 1
Katalog # STMC78067
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Human Recombinant TGF-beta 1
Human Recombinant TGF-beta 1

5 µG

CAS nummer:  
Price
1.721,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Add this high-quality human recombinant transforming growth factor beta 1 (TGF-β1) to your cell culture workflow. Its verified endotoxin level of ≤0,2 EU/μg, measured using the LAL method, allows confidence for use across multiple applications. A member of the TGF-β superfamily, TGF-β1 regulates diverse cellular phenotypes. TGF-β1 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins.

TGF-β1 has been used for:
hPSC culture maintenance:
Use TGF-β1 together with basic fibroblast growth factor (bFGF) to support the culture of undifferentiated human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells (Amit et al. Biol Reprod, 2004).
Exogenus cell signaling:
Regulate a variety of functions, including cell proliferation, differentiation, and metabolism (Massagué Nat Rev Mol Cell Biol, 2000; McDowell et al. Curr Biol, 1997).
Immune cell Homeostatis:
Modulate B cell responses and mediate immunosuppressive effects on T cells and neutrophils (Letterio and Roberts Annu Rev Immunol, 1998).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    5 µG
  • Cat. No.:
    STMC78067
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    TGF-beta 1
  • Renhed:
    ≥95%
  • Species:
    Human

Specifikationer