Orders LinkContactUsLinkComponent
Cell Lysis Buffer, Abcam
Cell Lysis Buffer, Abcam
Cell Lysis Buffer
Katalog # ABCAAB288311-100
Marketsource
Leverandør:   Abcam
Cell Lysis Buffer, Abcam
Katalog # ABCAAB288311-100
Leverandør:  Abcam
Marketsource
CAS nummer:  
Cell Lysis Buffer, Abcam
Cell Lysis Buffer
CAS nummer:  
Price
901,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
This product is manufactured by BioVision, an Abcam company and was previously called 1067 Cell Lysis Buffer.
  • Constituents: 1% Alcohol Ethoxylate, 0.88% Sodium chloride, 0.32% Tris HCl
Advarsel: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifikationer
Specifikationer
  • Beskrivelse:
    Cell Lysis Buffer
  • Cat. No.:
    ABCAAB288311-100
  • Opbevaringstemperatur:
    Store at +4°C.
Specifikationer