Masterflex® Fitting, PVDF, Tee, Hose Barb Reducer, 3/4" ID x 3/4" ID x 3/8" ID; 10/PK
Katalog # MFLX40615-51
Leverandør: Avantor Fluid Handling
Specifikationer
Se alle produktkonfigurationer- Catalog No:
- MFLX40615-51
- MFLX30401-58
- MFLX31702-61
- MFLX30401-60
- MFLX31702-57
- MFLX30401-55
- MFLX30401-59
- MFLX30401-56
- MFLX31208-48
- MFLX30401-54
- MFLX31208-46
- MFLX31208-47
- MFLX31208-44
- MFLX31208-45
- MFLX31208-41
- MFLX30401-63
- MFLX31702-47
- MFLX40615-48
- MFLX31702-45
- MFLX40614-36
- MFLX40615-50
- MFLX31702-62
- MFLX31208-42
- Model:
- 30401-58
- 31702-61
- 30401-60
- 31702-57
- 30401-55
- 30401-59
- 30401-56
- 31208-48
- 30401-54
- 31208-46
- 31208-47
- 31208-44
- 31208-45
- 31208-41
- 30401-63
- 31702-47
- 31702-45
- 31702-62
- 31208-42
- Orifice dimensions:
- 15,08 mm (0,59375")
- 0,34375"
- 0,25"
- 0,1875"
- 0,1875"
- 0,09375"
- 0,09375"
- 0,09375"
- 0,0625"
- 0,1875"
- 0,0625"
- 0,0625"
- 11,9 mm (0,46875")
- 0,34375"
- 0,0625"
- Sterilisering:
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Autoclave or ethylene oxide (EtO)
- Ethylene oxide (ETO) only
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) only
- Autoclave, ethylene oxide (ETO), or gamma
- Garanti:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Viser i øjeblikket
Specifikationer
Om denne vare
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Modhagestørrelse/slange int.Ø (port 1):3/4"
- Modhagestørrelse/slange int.Ø (port 2):3/4"
- Tilslutningstype:Barbed Reducer
- Flow pattern:T
- Beskrivelse:Fitting, PVDF, Tee, Hose Barb Reducer, 3/4" ID x 3/4" ID x 3/8" ID; 10/PK
- Environmentally Preferable:
- Orifice dimensions:15,08 mm (0,59375")
- Sterilisering:Autoclave or ethylene oxide (ETO)
- Pakn:10
- Barb size/tubing int.Ø (port 3):3/8"
- Fitting material:PVDF
Frequently Bought Together