Lids for plastic pallet boxes
Lid for pallet container, 1200×1000 mm, grå
Katalog # MULCADE1210
Leverandør: MULTIROIR
Gå til:
Product Details & Documents
Stackable HDPE lids for pallet boxes and containers. Suitable for storing foodstuffs.
Specifikationer
- Farve:grå
- Beskrivelse:Lid for pallet container, 1200×1000 mm
- Seq:02