Orders LinkContactUsLinkComponent
Micro soxhlet extraction apparatus, WHEATON®
Micro soxhlet extraction apparatus, WHEATON®
Soxhlet extraction apparatus, component parts, extraction thimble, glass 15×60 mm, 170-220 µm porosity, — ml
Katalog # WHEAW416788
Leverandør:   DWK Life Sciences
Micro soxhlet extraction apparatus, WHEATON®
Katalog # WHEAW416788
Leverandør:  DWK Life Sciences
Micro soxhlet extraction apparatus, WHEATON®
Micro soxhlet extraction apparatus, WHEATON®
Soxhlet extraction apparatus, component parts, extraction thimble, glass 15×60 mm, 170-220 µm porosity, — ml
Price
933,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.
Specifikationer
  • Beskrivelse:
    Soxhlet extraction apparatus, component parts, extraction thimble, glass 15×60 mm, 170-220 µm porosity
  • Seq:
    04
  • Kolbes volumen:
    — ml