Micro soxhlet extraction apparatus, WHEATON®
Soxhlet extraction apparatus, component parts, extraction thimble, glass 15×60 mm, 170-220 µm porosity, — ml
Katalog # WHEAW416788
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.
Specifikationer
- Beskrivelse:Soxhlet extraction apparatus, component parts, extraction thimble, glass 15×60 mm, 170-220 µm porosity
- Seq:04
- Kolbes volumen:— ml