Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumhydroxid solution 10%, AnalaR NORMAPUR®

HDPE bottle, 1 L
  85660.290
 
Natriumhydroxid solution 10%, AnalaR NORMAPUR®
  85660.290
 :  1310-73-2
Natriumhydroxid solution 10%, AnalaR NORMAPUR®
Natriumhydroxid solution 10%, AnalaR NORMAPUR®

HDPE bottle, 1 L

 :  1310-73-2
 
389,00 kr
   
1 * 1 L
 
 :  Caustic soda
  •  :1310-73-2
  •  :NaOH
  •  :MFCD00003548
  •  :40 g/mol
  •  :1,109 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :1824
  •  :8,II
 

 

  • Assay :
    10.0 - 10.5%
  • Heavy metals (as Pb):
    max. 5 ppm
  • Total nitrogen (N):
    max. 1 ppm
  • Carbonate (as Na₂CO₃):
    max. 1%
  • Chloride (Cl):
    max. 10 ppm
  • Phosphate (PO₄):
    max. 5 ppm
  • Silicate (as SiO₂):
    max. 10 ppm
  • Sulphate (SO₄):
    max. 5 ppm
  • Aluminium (Al):
    max. 5 ppm
  • Iron (Fe):
    max. 5 ppm

 

  • Pakning:
    HDPE bottle
  • Pakn:
    1 L