Masterflex® Sanitary Gaskets, Polymers, Avantor®
Sanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PK, Buna-N, 2", 2-1/2"
Catalog # MFLX30548-48
Supplier: Avantor Fluid Handling
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Catalog # MFLX30548-48
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Ensure a tight seal.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimise downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
Specifications
- Sanitary clamp size (Port 1):2"
- Beskrivelse:Sanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PK
- Seq:05
- Fitting material:Buna-N
- Sanitary gasket size:2-1/2"
- Cat. No.:MFLX30548-48
- Connection type:Sanitary
- Garanti:1 year