Masterflex® L/S® Easy-Load® Pump Head with Long-Shaft for Precision Tubing, 6-Roller, PARA Housing, SS Rotor
Katalog # MFLX07514-60
Leverandør: Avantor Fluid Handling
2 year warranty
Om denne vare
Save time with fast tubing changes and reduced maintenance.
- New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
- Rotor is constructed of stainless steel (SS) for enhanced durability
This L/S® Easy-Load® II Pump Head, when combined with a Masterflex® L/S® drive or compatible system, is designed to provide a simple, easy-to-use peristaltic pump system. The pump head accepts several different tubing sizes for a wide range of flow rates.
- Antal kanaler:1
- Pump series:L/S
- Beskrivelse:L/S® Easy-Load® Pump Head with Long-Shaft for Precision Tubing, 6-Roller, PARA Housing, SS Rotor
- Seq:03
- Environmentally Preferable:
- Model:07514-60
- Occlusion:Fixed
- Open-head sensor:No
- Tubing retention:Automatic
- Tubing sizes accepted:L/S® 13, L/S 14, L/S 16, L/S 25
- Garanti:2 years
- Pakn:1
- Ruller:6
- Housing material:Polyarylamide (PARA)
- Rotor material:Stainless Steel
Frequently Bought Together