Orders LinkContactUsLinkComponent
Dishes, crystallising
Dishes, crystallising
300 ml, 55 mm, 95 mm
Katalog # KAVA632411624095
Leverandør:   Kavalier
Dishes, crystallising
Katalog # KAVA632411624095
Leverandør:  Kavalier
Dishes, crystallising
Dishes, crystallising
300 ml, 55 mm, 95 mm
Price
2.210,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications.
Specifikationer
  • Udv.Ø:
    95 mm
  • Kapacitet:
    300 ml
  • Højde:
    55 mm
  • Seq:
    08