Hedland Flowmeter Accessories
AC Switch for spring-loaded in-line flowmeters, 0,5 A max, 115 VAC
Katalog # MFLX03231-80
Leverandør: Avantor Fluid Handling
Gå til:
Product Details & Documents
Make the most of your Hedland flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
Replacement switches and cables are suitable for use with Hedland spring-loaded in-line flowmeters or turbine flowmeters.
Specifikationer
- Beskrivelse:AC Switch for spring-loaded in-line flowmeters, 0,5 A max, 115 VAC
- Cat. No.:MFLX03231-80
- Garanti:1 year