- Catalog No:
- MFLX30627-71
- MFLX30623-77
- MFLX40613-05
- MFLX30623-73
- MFLX40624-27
- MFLX30627-58
- MFLX40613-01
- MFLX40623-64
- MFLX40627-56
- MFLX40623-56
- MFLX30623-58
- MFLX30613-33
- MFLX30623-71
- MFLX30627-73
- MFLX30623-60
- MFLX30623-67
- MFLX30627-79
- MFLX30627-60
- MFLX90009-27
- MFLX30623-79
- MFLX30627-77
- MFLX30627-67
- Model:
- 30627-71
- 30623-77
- 40613-05
- 30623-73
- 40624-27
- 30627-58
- 40613-01
- 40623-64
- 40627-56
- 40623-56
- 30623-58
- 30613-33
- 30623-71
- 30627-73
- 30623-60
- 30623-67
- 30627-79
- 30627-60
- 90009-27
- 30623-79
- 30627-77
- 30627-67
- Orifice dimensions:
- 0,125"
- 0,25"
- 0,1875"
- 0,46875"
- 0,46875"
- 0,125"
- 0,1875"
- 0,5625"
- 0,09375"
- 0,34375"
- 0,5625"
- 0,34375"
- 0,25"
- 0,09375"
- Sterilisering:
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene Oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene Oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene Oxide (ETO)
- Ethylene Oxide (ETO)
- Ethylene Oxide (ETO)
- Ethylene Oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Modhagestørrelse/slange int.Ø (port 1):1/4"
- Modhagestørrelse/slange int.Ø (port 2):1/4"
- Tilslutningstype:Barbed Union
- Flow pattern:T
- Beskrivelse:Fitting, HDPE, Tee, Hose Barb Union, 1/4" ID; 100/PK
- Seq:18
- Environmentally Preferable:
- Model:30627-71
- Orifice dimensions:0,125"
- Sterilisering:Ethylene oxide (ETO) only
- Pakn:100
- Barb size/tubing int.Ø (port 3):1/4"
- Fitting material:HDPE
Frequently Bought Together