Micropump Pump Head Adapter Kits
Series Q pump head to drive adapter kit
Katalog # MFLX07104-48
Leverandør: Avantor Fluid Handling
Gå til:
Product Details & Documents
Easily adapt your pump head to your drive with these adapter kits.
- Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.
Specifikationer
- Beskrivelse:Series Q pump head to drive adapter kit
- Cat. No.:MFLX07104-48
- Garanti:1 year