Masterflex® Fitting, Polypropylene, Straight, Hosebarb to Thread, 1/2" ID x 3/8" NPT(M); 10/PK
MFLX40610-43
CE certificate
- Catalog No:
- MFLX40610-43
- MFLX40625-61
- MFLX40625-41
- MFLX40625-51
- MFLX31407-16
- MFLX40610-46
- MFLX40610-50
- MFLX40625-28
- MFLX40610-53
- MFLX40625-45
- MFLX40610-47
- MFLX40610-48
- MFLX31407-13
- MFLX31407-14
- MFLX40610-51
- MFLX31407-12
- MFLX40610-52
- MFLX40610-37
- MFLX40610-49
- MFLX06365-42
- MFLX31407-19
- MFLX31407-20
- MFLX40621-61
- MFLX41517-27
- MFLX41517-47
- MFLX40621-45
- MFLX40621-51
- MFLX40640-51
- MFLX40640-50
- MFLX40640-46
- MFLX40640-44
- MFLX40640-43
- MFLX40640-42
- MFLX40640-41
- MFLX40640-38
- MFLX40621-11
- MFLX40621-28
- MFLX40621-41
- MFLX40621-22
- MFLX40621-25
- MFLX30615-51
- MFLX40610-42
- MFLX40610-40
- MFLX40610-41
- MFLX40640-39
- MFLX40640-45
- MFLX40610-44
- MFLX40640-36
- MFLX40610-45
- MFLX40640-37
- MFLX40621-69
- MFLX40610-38
- MFLX40610-39
- MFLX40610-36
- Model:
- 40610-43
- 40625-61
- 40625-41
- 40625-51
- 31407-16
- 40610-46
- 40610-50
- 40625-28
- 40610-53
- 40625-45
- 40610-47
- 40610-48
- 31407-13
- 31407-14
- 40610-51
- 31407-12
- 40610-52
- 40610-37
- 40610-49
- 06365-42
- 31407-19
- 31407-20
- 40621-61
- 41517-27
- 41517-47
- 40621-45
- 40621-51
- 40640-51
- 40640-50
- 40640-46
- 40640-44
- 40640-43
- 40640-42
- 40640-41
- 40640-38
- 40621-11
- 40621-28
- 40621-41
- 40621-22
- 40621-25
- 30615-51
- 40610-42
- 40610-40
- 40610-41
- 40640-39
- 40640-45
- 40610-44
- 40640-36
- 40610-45
- 40640-37
- 40621-69
- 40610-38
- 40610-39
- 40610-36
- No. of ports:
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- Orifice dimensions:
- 8,73 mm (0,34375")
- 11,9 mm (0,46875")
- 8,73 mm (0,34375")
- 14,29 mm (0,5625")
- 8,73 mm (0,34375")
- 3,175 mm (0,125")
- 6,35 mm (0,25")
- 4,763 mm (0,1875")
- 11,9 mm (0,46875")
- 3,175 mm (0,125")
- 9,53 mm (0,375")
- 6,35 mm (0,25")
- 6,35 mm (0,25")
- 8,73 mm (0,34375")
- 19,84 mm (0,78125")
- 11,9 mm (0,46875")
- 17,46 mm (0,6875")
- 3,175 mm (0,125")
- 14,29 mm (0,5625")
- 8,73 mm (0,34375")
- 3,175 mm (0,125")
- 8,73 mm (0,34375")
- 6,35 mm (0,25")
- 3,175 mm (0,125")
- 3,175 mm (0,125")
- 6,35 mm (0,25")
- 8,73 mm (0,34375")
- 3,175 mm (0,125")
- 6,35 mm (0,25")
- 6,35 mm (0,25")
- 3,175 mm (0,125")
- 3,175 mm (0,125")
- 6,35 mm (0,25")
- 6,35 mm (0,25")
- 3,175 mm (0,125")
- 6,35 mm (0,25")
- 3,175 mm (0,125")
- Sterilisering:
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Garanti:
- Out of box
- 1 year
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- 1 year
- 1 year
- Out of box
- 1 year
- Out of box
- Out of box
- 1 year
- 1 year
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- 1 year
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Modhagestørrelse/slange int.Ø (port 2):1/2"
- Tilslutningstype:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):3/8"
- Beskrivelse:Fitting, Polypropylene, Straight, Hosebarb to Thread, 1/2" ID x 3/8" NPT(M); 10/PK
- Seq:24
- Environmentally Preferable:
- Model:40610-43
- No. of ports:2
- Orifice dimensions:8,73 mm (0,34375")
- Sterilisering:Ethylene oxide (ETO) or gamma
- Garanti:Out of box
- Pakn:10
- Fitting material:Polypropylene
Frequently Bought Together