Orders LinkContactUsLinkComponent
BRUTE® Cleaning Accessories
BRUTE® Cleaning Accessories
BRUTE® caddy bag, vinyl, black
Catalog # 129-2196
BRUTE® Cleaning Accessories
Catalog # 129-2196
BRUTE® Cleaning Accessories
BRUTE® Cleaning Accessories
BRUTE® caddy bag, vinyl, black
Price
633,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Turn Rubbermaid Commercial BRUTE® waste containers into a cleaning cart with storage and transport for spray bottles, wet-floor signs, lobby dust pan, brushes, liners, gloves, and other cleaning supplies.
  • Rim caddy snaps securely onto the rim of 167 litre BRUTE® waste containers
  • Caddy bag fits 121 and 167 litre BRUTE® waste containers
The BRUTE® rim caddy measures 825×673×171 mm. BRUTE® caddy bags measure 387×286×44 mm.
Specifications
  • Beskrivelse:
    BRUTE® caddy bag, vinyl, black
  • Seq:
    02