Orders LinkContactUsLinkComponent
HYGEN™ Quick-Connect Flexible Dusting Wand
Flexible dusting wand, hvid, Mikrofiber
  RUBBFGQ85200WH00
HYGEN™ Quick-Connect Flexible Dusting Wand
  RUBBFGQ85200WH00
HYGEN™ Quick-Connect Flexible Dusting Wand
HYGEN™ Quick-Connect Flexible Dusting Wand
Flexible dusting wand, hvid, Mikrofiber
337,00 kr
1 * 1 Items
This HYGEN™ flexible duster is perfect for removing dust from harder-to-reach areas and is efficient for use even on irregular surfaces. The removable microfibre sleeve is extremely hard-wearing and will stand up to washing and drying at 95 °C up to 500 times, offering great value for money.
  • Easily bends and conforms to irregular shapes, allowing users to clean crown molding, cabinet tops, and other areas
  • Quick-Connect handle allows Flexi-Wand to be used with any of the telescoping handles
  • Washable duster sleeve slides on and off for easy replacement
  • Measures 28¹/₃×1¹/₈"
  • Can be used dry or damp
The Rubbermaid Commercial HYGEN™ microfibre quick-connect flexi-wand and duster connects with a telescoping handle (sold separately) to easily clean vents, furniture, bed covers, uniquely shaped fixtures and more.
  • Farve:
    hvid
  • Beskrivelse:
    Flexible dusting wand
  • Materiale:
    Mikrofiber
  • Seq:
    1
Complementary Products
  •  
    Beskrivelse
    Materiale
    Længde
    Dimensioner
    Farve
  • RUBBFGQ75500YL00
    Large extension handle
    aluminium
    182,9 - 548,6 cm
    5,3 cm Ø
    gul
    1 * 1 Items - 312,00 kr
  • RUBBFGQ74500YL00
    Short extension handle
    aluminium
    50,8 - 101,6 cm
    5,1 cm Ø
    gul
    1 * 1 Items - 233,00 kr