Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HYGEN™ Quick-Connect Handles

HYGEN™ Quick-Connect Handles

Large extension handle, gul, 5,3 cm Ø, 182,9 - 548,6 cm, aluminium
Katalog # RUBBFGQ75500YL00
HYGEN™ Quick-Connect Handles
Katalog # RUBBFGQ75500YL00
HYGEN™ Quick-Connect Handles
HYGEN™ Quick-Connect Handles

Large extension handle, gul, 5,3 cm Ø, 182,9 - 548,6 cm, aluminium

Price
312,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

The HYGEN™ Quick-Connect handles and poles make cleaning more efficient in every area of the facility. The unique connection mechanism allows for easy, time-saving tool exchange.

  • Lightweight aluminum construction for improved productivity is safe for MRI rooms
  • One-step connecting mechanism speeds mop assembly and changeover

The user-friendly grip makes each handle more comfortable to use.

Specifikationer
  • Farve:
    gul
  • Beskrivelse:
    Large extension handle
  • Dimensioner:
    5,3 cm Ø
  • Længde:
    182,9 - 548,6 cm
  • Materiale:
    aluminium
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.