Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

HYGEN™ Quick-Connect Handles

Short extension handle, gul, 5,1 cm Ø, 50,8 - 101,6 cm, aluminium
Katalog # RUBBFGQ74500YL00
HYGEN™ Quick-Connect Handles
Katalog # RUBBFGQ74500YL00
HYGEN™ Quick-Connect Handles
HYGEN™ Quick-Connect Handles

Short extension handle, gul, 5,1 cm Ø, 50,8 - 101,6 cm, aluminium

Price
233,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

The HYGEN™ Quick-Connect handles and poles make cleaning more efficient in every area of the facility. The unique connection mechanism allows for easy, time-saving tool exchange.

  • Lightweight aluminum construction for improved productivity is safe for MRI rooms
  • One-step connecting mechanism speeds mop assembly and changeover

The user-friendly grip makes each handle more comfortable to use.

Specifikationer
  • Farve:
    gul
  • Beskrivelse:
    Short extension handle
  • Dimensioner:
    5,1 cm Ø
  • Længde:
    50,8 - 101,6 cm
  • Materiale:
    aluminium
  • Seq:
    01