Sterile goods baskets
Sterile goods baskets, stackable with access, 600×300×300 mm
Katalog # TEKMPBK0600/300/A
Leverandør: TEKNOMEK
Gå til:
Product Details & Documents
High-quality electro polished 304 grade stainless steel baskets, with a sterile design for maximum hygiene.
- Available with front access allows for easy access of stored goods
- Stronger welds for extra weight capacity and rigidity
- Wire structure allows optimal airflow
- Modular system stackable to maximise space saving
Fully welded in 304 grade stainless steel for maximum durability and corrosion resistance, it has an electro polished finish for ease of cleaning and to prevent build-up of residue. Available with sterile goods divider.
Specifikationer
- Beskrivelse:Sterile goods baskets, stackable with access
- Seq:06
- B×D×H:600×300×300 mm