Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Stainless steel scraper

230 mm, 75 mm
Katalog # 216-2503
Leverandør:   BURKLE
Stainless steel scraper
Katalog # 216-2503
Leverandør:  BURKLE
Stainless steel scraper
Stainless steel scraper

230 mm, 75 mm

Price
719,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

The high-quality scraper is suitable for scraping off, smoothing, cleaning, levelling out and also for removing strongly adhesive, sticky substances or dried up media. The edge of the corrosion-resistant stainless steel scraper is ideal for blending and portioning viscous media.

  • High-grade stainless steel V4A (1.4404), for ideal chemical resistance
  • Extra sturdy design guarantees long service life
  • Sterilisable, autoclavable

The smooth, seamless and sturdy surface ensures cleaning without difficulty and meets hygienic requirements.

The scraper makes a suitable implement for use in laboratories, production and clean rooms. It is ideal for use under rugged conditions in pharmaceuticals, chemistry, cosmetics, animal feed and food & beverage.

Specifikationer
  • Aftørringsområde ca.:
    75 mm
  • Længde:
    230 mm
  • Seq:
    01