Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro spatula, round, -, 150 mm, 3 mm
Katalog # RSGA80.530.003
Leverandør:   RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
Katalog # RSGA80.530.003
Leverandør:  RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro spatula, round, -, 150 mm, 3 mm

Price
kr148.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Made from stainless steel 1.4301.

  • Rust-proof
  • Highly resistant against chemicals
  • With 6 edged grip for better handling
  • Autoclavable
Specifikationer
  • Steril:
    -
  • Beskrivelse:
    Micro spatula, round
  • Længde:
    150 mm
  • Seq:
    04
  • Bladbredde:
    3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.