Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro powder spatula, -, 150 mm, 4 mm
  RSGA80.532.004
 :   RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
  RSGA80.532.004
 :  RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro powder spatula, -, 150 mm, 4 mm

 
154,00 kr
   
1 * 1 Items
 

Made from stainless steel 1.4301.

  • Rust-proof
  • Highly resistant against chemicals
  • With 6 edged grip for better handling
  • Autoclavable
 
  • Steril:
    -
  • Beskrivelse:
    Micro powder spatula
  • Længde:
    150 mm
  • Seq:
    02
  • Bladbredde:
    4 mm