Orders LinkContactUsLinkComponent
Micro-Spatulas and Micro-Spoon, RSG Solingen
Micro spatula, straigth, -, 150 mm, 3 mm
  RSGA80.531.003
 :   RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
  RSGA80.531.003
 :  RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
Micro-Spatulas and Micro-Spoon, RSG Solingen
Micro spatula, straigth, -, 150 mm, 3 mm
148,00 kr
1 * 1 Items
Made from stainless steel 1.4301.
  • Rust-proof
  • Highly resistant against chemicals
  • With 6 edged grip for better handling
  • Autoclavable
  • Steril:
    -
  • Beskrivelse:
    Micro spatula, straigth
  • Længde:
    150 mm
  • Seq:
    06
  • Bladbredde:
    3 mm