Coupling body plugs
Coupling body plug, with class VI silicone O-ring, MPX series, Polykarbonat
Catalog # CPCCMPX30003M
Supplier: CPC COLDER
Gå til:
Product Details & Documents
Class VI coupling body plug with Class VI silicone O-ring.
Documents
Specifikationer
- Beskrivelse:Coupling body plug, with class VI silicone O-ring, MPX series
- Materiale:Polykarbonat
- Seq:03