Re-Usable Cleaning Coupons
Polypropylene, 100×100 mm
Katalog # SAMP8644PVC-100
Leverandør: Sampling Systems
Gå til:
Product Details & Documents
Cleaning coupons are great for validating a cleaning regime.
- All popular sizes available
- Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
Specifikationer
- Beskrivelse:Polypropylene
- L×B:100×100 mm
- Seq:12