Orders LinkContactUsLinkComponent
Wash troughs
Wash troughs
Infra-red mains operated sensor five station wash trough, wall mounted, fully welded 304-grade stainless steel, 2505W×380D×525H mm, 2500×123×283 mm
Katalog # TEKMWTN050EXSEN
Leverandør:   TEKNOMEK
Wash troughs
Katalog # TEKMWTN050EXSEN
Leverandør:  TEKNOMEK
Wash troughs
Wash troughs
Infra-red mains operated sensor five station wash trough, wall mounted, fully welded 304-grade stainless steel, 2505W×380D×525H mm, 2500×123×283 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienic standards.
Bestillingsoplysninger: Optional skirt (to be ordered alongside the wash trough as cannot be retrofitted).
Specifikationer
  • Beskrivelse:
    Infra-red mains operated sensor five station wash trough, wall mounted, fully welded 304-grade stainless steel, 2505W×380D×525H mm
  • Seq:
    26
  • B×D×H indv.:
    2500×123×283 mm