Add-on storage units
Table add-on 1 drawer with lock, 452×516×152 mm
Katalog # TEKMTCA0100
Leverandør: TEKNOMEK
Gå til:
Product Details & Documents
304 grade stainless steel add on unit for rear railed workbench is perfect for adding additional storage. Cannot be sold singly.
- Fully welded 304-grade stainless steel
- Brushed dull polish finish
Supplied fitted to workbench when ordered together. Choice of one drawer, two drawers, three drawers or cupboard unit, for our rear railed workbenches. Enables the user to create a workstation to suit their individual requirements.
Specifikationer
- Beskrivelse:Table add-on 1 drawer with lock
- Seq:03
- B×D×H:452×516×152 mm