Orders LinkContactUsLinkComponent
Basket wall supports
Basket wall supports, 4 tier, height 1600 mm
Catalog # TEKMPWB0010/4
Supplier:   TEKNOMEK
Basket wall supports
Catalog # TEKMPWB0010/4
Supplier:  TEKNOMEK
Basket wall supports
Basket wall supports, 4 tier, height 1600 mm
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
The wall mounted supports can be used in conjunction with our modular baskets.
  • Additional brackets suitable for our modular baskets
  • Material: 304-grade stainless steel
They are suitable for use with our modular baskets giving a range of storage options to suit your needs and come in a three and four tier option. They are priced per pair.
Specifications
  • Beskrivelse:
    Basket wall supports, 4 tier, height 1600 mm
  • Seq:
    02