Basket wall supports
Basket wall supports, 3 tier, height 1300 mm
Katalog # TEKMPWB0010/3
Leverandør: TEKNOMEK
Gå til:
Product Details & Documents
The wall mounted supports can be used in conjunction with our modular baskets.
- Additional brackets suitable for our modular baskets
- Material: 304-grade stainless steel
They are suitable for use with our modular baskets giving a range of storage options to suit your needs and come in a three and four tier option. They are priced per pair.
Specifikationer
- Beskrivelse:Basket wall supports, 3 tier, height 1300 mm
- Seq:01