Cryogenic markers, SCIENCE-MARKER™
Cryogenic permanent marker, Shipping weight 0,113 kg, 15,9×101,6×168,3 mm, grøn
Catalog # GAINSM-1G-6
Supplier: GA
Jump to:
Product Details & Documents
Cryogenic permanent marker with fine tip.
- Alcohol and water resistant
- Compatible with Lab-TAG™ labels
Specifications
- Farve:grøn
- Beskrivelse:Cryogenic permanent marker, Shipping weight 0,113 kg, 15,9×101,6×168,3 mm
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



