Floor cleaners, SURE®
Floor cleaner, 2×5 L
Catalog # 141-1572
Supplier: Diversey
Floor cleaners, SURE®
Catalog # 141-1572
Supplier: Diversey
Jump to:
Product Details & Documents
Concentrated, low foaming daily cleaner for all water resistant hard floors. Recommended for all cleaning methods, including bucket and scrubber drier applications.
- Concentrated formulation gives excellent economy of use
- No residues on the floor and no build-up
- Low foaming effective cleaning
- No fragrance or dye
- Contains plant-based ingredients from renewable sources
- Decomposed by natural processes
- No environmental hazard classifications, no safety warnings for user
SURE® floor cleaner is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
: OECD 301B.
EU-Ecolabel (EU: NL/020/045).
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.
Documents
TDS Diversey SURE Floor Cleaner
Datasheet
Specifications
- Beskrivelse:Floor cleaner, 2×5 L
- Seq:02
- Cat. No.:141-1572
- Environmentally Preferable:Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage