Orders LinkContactUsLinkComponent
Grill cleaners, SURE®
Grill cleaner, 2×5 L bottle
  DIVY100894371
   Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage
 :   Diversey
Grill cleaners, SURE®
  DIVY100894371
 :  Diversey
   Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage
Grill cleaners, SURE®
Grill cleaners, SURE®
Grill cleaner, 2×5 L bottle
604,00 kr
1 * 2 Fl
Detergent suitable for daily cleaning of heavily soiled surfaces, such as ovens and grills.
  • Specially developed for daily cleaning
  • Viscous product has excellent adherence on vertical surfaces
  • Ease of rinse
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications
SURE® grill cleaner is part of a unique range of plant-based, Cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
 : OECD 301B test
Cradle to cradle certification at gold level
  • Beskrivelse:
    Grill cleaner
  • Seq:
    02
  • Pakket:
    2×5 L bottle
  • Cat. No.:
    DIVY100894371
  • Environmentally Preferable:
    Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage