Grill cleaners, SURE®
Grill cleaner, 6×0,75 L bottle
Katalog # DIVY100943914
Leverandør: Diversey
Grill cleaners, SURE®
Katalog # DIVY100943914
Leverandør: Diversey
Gå til:
Product Details & Documents
Detergent suitable for daily cleaning of heavily soiled surfaces, such as ovens and grills.
- Specially developed for daily cleaning
- Viscous product has excellent adherence on vertical surfaces
- Ease of rinse
- No fragrance or dye
- Contains plant-based ingredients from renewable sources
- Decomposed by natural processes
- No environmental hazard classifications
SURE® grill cleaner is part of a unique range of plant-based, Cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test
Cradle to cradle certification at gold level
Documents
TDS Diversey SURE Grill Cleaner
Datablad
Specifikationer
- Beskrivelse:Grill cleaner
- Seq:01
- Pakket:6×0,75 L bottle
- Cat. No.:DIVY100943914
- Environmentally Preferable:Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage