Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Interior and surface cleaners, SURE®

Interior and surface cleaners, SURE®

Interior and surface cleaner, 2×5 L
Catalog # 115-2961
   Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage
Supplier:   Diversey
Interior and surface cleaners, SURE®
Catalog # 115-2961
Supplier:  Diversey
   Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage
Interior and surface cleaners, SURE®
Interior and surface cleaners, SURE®

Interior and surface cleaner, 2×5 L

Price
406,00 kr
Log in for availability
1 * 2 Fl
Product Details & Documents

Concentrated all-purpose detergent for cleaning of hard surfaces, such as floors, walls, doors, ceilings and general equipment. Can also be used in food premises.

  • Suitable for cleaning of all water resistant hard surfaces, including aluminium and other soft metals
  • Effective in all water conditions
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for user
  • Concentrated formulation gives excellent economy of use
  • Fast acting, fast drying

SURE® interior and surface cleaner is part of a unique range of plant-based, 100% biodegradable cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.

Certifications: EU-Ecolabel (EU:NL/020/045).
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.

Specifications
  • Beskrivelse:
    Interior and surface cleaner, 2×5 L
  • Seq:
    02
  • Cat. No.:
    115-2961
  • Environmentally Preferable:
    Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage