Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Snap Ring Vials ND11

VWR®, Snap Ring Vials ND11

Micro-vials, 0,30 ml, Conical bottom, self-standing, 11,6×32 mm, TPX, transparent, No, Snap, ND11
Catalog # 548-0451A
Supplier:   VWR Collection
VWR®, Snap Ring Vials ND11
Catalog # 548-0451A
Supplier:  VWR Collection
VWR®, Snap Ring Vials ND11
VWR®, Snap Ring Vials ND11

Micro-vials, 0,30 ml, Conical bottom, self-standing, 11,6×32 mm, TPX, transparent, No, Snap, ND11

Price
3.670,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Snap ring ND11 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyethylene seal styles and septa materials.

  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Wide opening

Snap ring ND11 vials constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear version with a flat bottom to ensure compatibility with inserts. Multiple cap colours mostly available with assembled septa are engineered to prevent sample contamination while providing consistent, secure seals. Furthermore, snap ring vials ND11 are available in amber and transparent plastic version.

Specifications
  • Kasket inkluderet:
    No
  • Cap størrelse:
    ND11
  • Diameter×H:
    11,6×32 mm
  • Kapacitet:
    0,30 ml
  • Farve:
    transparent
  • Beskrivelse:
    Micro-vials
  • Materiale:
    TPX
  • Låg type:
    Snap
  • Grundlag:
    Conical bottom, self-standing
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.