Cold trap, type KF54V
Cold trap, KF 54V-K25-Z-18C, stainless steel, with 200 ml condensate capacity and 1600 ml coolent cap
Katalog # KGWA17112
Leverandør: KGW
Gå til:
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents.
- Easy to clean
- No stand material for holding the cold trap
- Protective casing of Dewar flask
Specifikationer
- Beskrivelse:Cold trap, KF 54V-K25-Z-18C, stainless steel, with 200 ml condensate capacity and 1600 ml coolent cap
- Seq:04