Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Assistent® Brushes with Cotton Tip Bristles

Assistent® Brushes with Cotton Tip Bristles

Bomuld, 20 mm, Galvanised wire, 300 mm
Katalog # HECH42760010
Assistent® Brushes with Cotton Tip Bristles
Katalog # HECH42760010
Assistent® Brushes with Cotton Tip Bristles
Assistent® Brushes with Cotton Tip Bristles

Bomuld, 20 mm, Galvanised wire, 300 mm

Price
363,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

Brushes, light bristles, with cotton tip.

Specifikationer
  • Børstemateriale:
    Bomuld
  • Børstehoved Ø:
    20 mm
  • Handle material:
    Galvanised wire
  • Seq:
    08
  • Samlet længde:
    300 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.