Orders LinkContactUsLinkComponent
Sprøjtefiltre, J.T.Baker®
Sprøjtefiltre, J.T.Baker®
30 mm, 0,22 µm, Nylon, Bulk
Katalog # SF01-72
Leverandør:   Avantor
Sprøjtefiltre, J.T.Baker®
Katalog # SF01-72
Leverandør:  Avantor
Sprøjtefiltre, J.T.Baker®
Sprøjtefiltre, J.T.Baker®
30 mm, 0,22 µm, Nylon, Bulk
Price
kr10,370.00
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Disse sprøjtefiltre er specielt designet til kromatografiprøveforberedelse og optimeret til at give de mest ensartede resultater med færre ekstraherbare materialer.
  • HPLC certificeret
  • Biosikker i henhold til klasse VI plastiktest
  • Husmateriale af polypropylen
  • Ikke steril
  • Hvert filter har et hun-luer-lock-indløb og et han-slip-luer-udløb
  • Stor porestørrelse 1,0 µm for høj gennemstrømning og hurtige flowhastigheder
Available with PTFE (hydrophilic or hydrophobic), nylon, PES, RC membrane and GF (glass fibre):
- Water wettable hydrophilic PTFE offers excellent chemical resistance to both common aqueous and aggressive organic solvents, most of the acids and alkalis
- Hydrophobic PTFE resistant to the majority of solvents as well as strong acids and bases, also suitable for gas filtration
- Nylon provides a wide range of chemical compatibility, suitable for filtration of either aqueous or organic solvents. It is a natural hydrophilic membrane that can be used in a broad pH range
- PES offers lower protein binding as well as higher flow rates
- Regenerated cellulose exhibits low protein binding and wide chemical resistance
- Built in glass fibre (100% binder-free borosilicate glass fibre) pre-filter double-layer type is specially for high particle loading, ideal for difficult to filter liquids
Certifications: Produceret under ISO 9001:2015 kvalitetsstyring.
Specifikationer
  • Ø:
    30 mm
  • Membran:
    Nylon
  • Pakket:
    Bulk
  • Porestørrelse:
    0,22 µm
  • Seq:
    34