Sprøjtefiltre, J.T.Baker®
Product Details & Documents
Disse sprøjtefiltre er specielt designet til kromatografiprøveforberedelse og optimeret til at give de mest ensartede resultater med færre ekstraherbare materialer.
- HPLC certificeret
- Biosikker i henhold til klasse VI plastiktest
- Husmateriale af polypropylen
- Ikke steril
- Hvert filter har et hun-luer-lock-indløb og et han-slip-luer-udløb
- Stor porestørrelse 1,0 µm for høj gennemstrømning og hurtige flowhastigheder
Available with PTFE (hydrophilic or hydrophobic), nylon, PES, RC membrane and GF (glass fibre):
- Water wettable hydrophilic PTFE offers excellent chemical resistance to both common aqueous and aggressive organic solvents, most of the acids and alkalis
- Hydrophobic PTFE resistant to the majority of solvents as well as strong acids and bases, also suitable for gas filtration
- Nylon provides a wide range of chemical compatibility, suitable for filtration of either aqueous or organic solvents. It is a natural hydrophilic membrane that can be used in a broad pH range
- PES offers lower protein binding as well as higher flow rates
- Regenerated cellulose exhibits low protein binding and wide chemical resistance
- Built in glass fibre (100% binder-free borosilicate glass fibre) pre-filter double-layer type is specially for high particle loading, ideal for difficult to filter liquids
Certifications: Produceret under ISO 9001:2015 kvalitetsstyring.
Documents
Certifikater
Søg efter partinummerAvantor JTBaker Syringe Filter Chemical Compatibility J.T.Baker® Avantor Syringe Filters
Product Literature
BAKRSF01-16_334016023.pdf BAKRSF01-18_328119295.pdf BAKRSF01-24_329123078.pdf BAKRSF01-32_321118292.pdf BAKRSF01-38_313059048.pdf
J.T.Baker® Syringe Filters, Avantor®
Application Notes
Specifikationer
- Ø:30 mm
- Membran:Hydrofil PTFE
- Pakket:2 glas med100
- Porestørrelse:0,45 µm
- Seq:19