Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Brefeldin A

Brefeldin A

Bottle, 1 g
Katalog # APOSBIB3042-1G
Giftig
Leverandør:   Apollo Scientific
Brefeldin A
Katalog # APOSBIB3042-1G
Leverandør:  Apollo Scientific
CAS nummer:  20350-15-6
Brefeldin A

Bottle, 1 g

CAS nummer:  20350-15-6
Price
16.470,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

BFA is a macrolide antiobiotic. Disrupts mechanism of Golgi apparatus.

Synonymer:  (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
  • CAS nummer:20350-15-6
  • Formel:C₁₆H₂₄O₄
  • MDL nummer:MFCD00083258
  • Molecular Weight:280.4 g/mol
  • Smeltende Pt:200…205 °C
  • Opbevaringstemperatur:Køleskab
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    1 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.