Cooling Blocks, Passive Cooler
Cooling block, passive cooler, PP/aluminium, 384 well PCR plates
Katalog # BRND709514
Leverandør: Brand
Gå til:
Product Details & Documents
Keep your samples and media cool for up to three hours with these Cooling Blocks from BRAND.
ANSI/SLAS format for use in pipetting robotic systems such as the Liquid Handling Station
- Cools microcentrifuge tubes and PCR consumables
All well positions are cooled homogenously. The Cooling Blocks work as a passive heat sink which makes them quick and easy to use without an additional power supply.
Documents
Cooling Blocks.pdf
Ansøgningsnoter
Manual Cooling Block.pdf
Manuel
TDS Liquid Handling Station.pdf
Datablad
Specifikationer
- Beskrivelse:Cooling block, passive cooler, PP/aluminium
- Seq:05
- Til:384 well PCR plates