Orders LinkContactUsLinkComponent
Cooling Blocks, Passive Cooler
Cooling Blocks, Passive Cooler
Cooling block, passive cooler, PP/aluminium, 384 well PCR plates
Katalog # BRND709514
Leverandør:   Brand
Cooling Blocks, Passive Cooler
Katalog # BRND709514
Leverandør:  Brand
Cooling Blocks, Passive Cooler
Cooling Blocks, Passive Cooler
Cooling block, passive cooler, PP/aluminium, 384 well PCR plates
Price
9.420,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Keep your samples and media cool for up to three hours with these Cooling Blocks from BRAND.
  • ANSI/SLAS format for use in pipetting robotic systems such as the Liquid Handling Station
  • Cools microcentrifuge tubes and PCR consumables
All well positions are cooled homogenously. The Cooling Blocks work as a passive heat sink which makes them quick and easy to use without an additional power supply.
Specifikationer
  • Beskrivelse:
    Cooling block, passive cooler, PP/aluminium
  • Seq:
    05
  • Til:
    384 well PCR plates