Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-(4-Hydroxyphenyl)ethanol ≥98%

2-(4-Hydroxyphenyl)ethanol ≥98%

250 g
Katalog # CAYM27600-250
Irriterende
Leverandør:   Cayman Chemical
2-(4-Hydroxyphenyl)ethanol ≥98%
Katalog # CAYM27600-250
Leverandør:  Cayman Chemical
CAS nummer:  501-94-0
2-(4-Hydroxyphenyl)ethanol ≥98%

250 g

CAS nummer:  501-94-0
Price
2.240,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Tyrosol is a phenol that has been found in olives and olive oil and has diverse biological activities.

 : This product is not for human or veterinary use.

Synonymer:  NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
  • CAS nummer:501-94-0
  • Formel:C₈H₁₀O₂
  • MDL nummer:MFCD00002902
  • Molecular Weight:138.17 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    250 g

Specifikationer