(-)-Indolactam V ≥97%, solid
500 µG
Katalog # CAYM14647-500
Leverandør: Cayman Chemical
(-)-Indolactam V ≥97%, solid
Katalog # CAYM14647-500
Leverandør: Cayman Chemical
CAS nummer: 90365-57-4
Gå til:
Product Details & Documents
(−)-Indolactam V is a tumor-promoting teleocidin compound that activates protein kinase C (PKC). It binds to PKC regulatory domains of mouse skin PKCη and rat brain PKCγ with Ki values of 3,4 nM and 1 μM, respectively. At 1 μM, (−)-indolactam V induces differentiation of human embryonic stem cells into pancreatic progenitors through activation of PKC signaling.
- CAS nummer:90365-57-4
- Formel:C₁₇H₂₃N₃O₂
- Molecular Weight:301.39 g/mol
Specifikationer
Specifikationstestresultater
- Formal name:(2S,5S)-1,2,4,5,6,8-hexahydro-5-(hydroxymethyl)-1-methyl-2-(1-methylethyl)-3H-pyrrolo[4,3,2-gh]-1,4-benzodiazonin-3-one
- CAS number:90365-57-4
- Molecular formula:C17H23N3O2
- Formula weight:301,4
- Purity:≥97%
- Formulation:A white to off-white solid
- λmax:203, 229 and 301 nm
- Storage:4 °C
- Stability:≥ 2 years
Specifikationer
- Pakn:500 µG
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



