Glycin ≥98%, GPR RECTAPUR®
Plastic container, 25 kg
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS nummer:56-40-6
- Formel:NH₂CH₂COOH
- MDL nummer:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Kogende Pt:181 °C (1013 hPa)
- Smeltende Pt:83 °C
- Tæthed:1,64 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-272-2
Specifikationer
Specifikationstestresultater
- Assay:Min. 98 %
- Ignition residue (SO4):Max. 0.2 %
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.02 %
Specifikationer
- Pakning:Plastic container
- Pakn:25 kg



