Celite® 512, GPR RECTAPUR®
PP bucket, 1 kg
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Calcined Diatomaceous Earth Filter Aid.
Celite® is used as a filtering aid or as adsorbent in column chromatography.
- CAS nummer:91053-39-3
- Formel:SiO₂
- Molecular Weight:60.08 g/mol
Documents
Certifikater
Søg efter partinummerP20_VWRC22555.290_SPEC_EN.pdf spec_VWRC22555.290 spec_VWRC22555.360 spec_VWRC22555.460
Produktspecifikation
VWR® for Drying and Filtration
Produktlitteratur
Specifikationer
Specifikationstestresultater
- pH (25 °C; 10%):8,5 - 11,0
- Wet density:≤ 370 g/l
- Loss on ignition (1000 °C):≤ 0,4%
- Particle size (< 75µm):≥ 90%
Specifikationer
- Pakning:PP bucket
- Pakn:1 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



