Orders LinkContactUsLinkComponent
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons
Silicone, 50×50 mm
  SAMP8644SIL-50
Re-Usable Cleaning Coupons
  SAMP8644SIL-50
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons
Silicone, 50×50 mm
1.650,00 kr
1 * 1 Items
Cleaning coupons are great for validating a cleaning regime.
  • All popular sizes available
  • Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
  • Beskrivelse:
    Silicone
  • L×B:
    50×50 mm
  • Seq:
    05