Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
X-Gal ≥98%

X-Gal ≥98%

Bottle, 10 g
Catalog # APOSBIMB1001-10G
Irritant
Supplier:   Apollo Scientific
X-Gal ≥98%
Catalog # APOSBIMB1001-10G
Supplier:  Apollo Scientific
CAS Number:  7240-90-6
X-Gal ≥98%

Bottle, 10 g

CAS Number:  7240-90-6
Price
1.010,00 kr
Log in for availability
1 * 10 g
Product Details & Documents

X-Gal, a chromogenic substrate of beta-Galactosidase. Used in conjunction with IPTG for detection of beta-Galactosidase activity in bacterial colonies in a colorimetric assay.

Synonyms:  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  • CAS Number:7240-90-6
  • Formula:C₁₄H₁₅BrClNO₆
  • MDL Number:MFCD00005666
  • Molecular Weight:408.63 g/mol
  • Melting Pt:230 °C
  • Storage Temperature:Fryser
  • EINECS:230-640-8
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.